Assay Details
Amplicon Length: | 84 |
Chromosome Location: | Chr. 15 - 33613697 - 33613697 [+] on 33613697 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Rat | rno-miR-208a-3p | MIMAT0000880 |
Rat | rno-miR-208a-5p | MIMAT0017155 |
Stem-loop Details
Rat
