Assay Details
Gene Family ID: | MIPF0000178, mir-208 |
Mature miRNA Sequence: | GAGCUUUUGGCCCGGGUUAUAC |
miRBase Details:
Species | miRBase ID | miRBase Accession Number |
---|---|---|
Human | hsa-miR-208a-5p | MIMAT0026474 |
Mouse | mmu-miR-208a-5p | MIMAT0017014 |
Rat | rno-miR-208a-5p | MIMAT0017155 |
Dasypus novemcinctus | dno-miR-208a-5p | MIMAT0047750 |
Guinea pig | cpo-miR-208a-5p | MIMAT0047105 |
Pteropus alecto | pal-miR-208a-5p | MIMAT0040025 |
View More Rows | ||
Rhesus monkey | mml-miR-208a-5p | MIMAT0026844 |
Stem-loop Details
Human
Mouse
Rat
Dasypus novemcinctus
Guinea pig
Pteropus alecto
Rhesus monkey
