Assay Details
Gene Family ID: | MIPF0000178, mir-208 |
Mature miRNA Sequence: | GAGCUUUUGGCCCGGGUUAUAC |
Mature miRNA Details:
Species | miRBase ID | miRBase Accession Number | |
---|---|---|---|
Human | hsa-miR-208a-5p | MIMAT0026474 | |
Mouse | mmu-miR-208a-5p | MIMAT0017014 | |
Rat | rno-miR-208a-5p | MIMAT0017155 | |
Dasypus novemcinctus | dno-miR-208a-5p | MIMAT0047750 | |
Guinea pig | cpo-miR-208a-5p | MIMAT0047105 | |
Pteropus alecto | pal-miR-208a-5p | MIMAT0040025 | |
View More Rows | |||
Rhesus monkey | mml-miR-208a-5p | MIMAT0026844 |
Stem-loop Details
Human
Mouse
Rat
Dasypus novemcinctus
Guinea pig
Pteropus alecto
Rhesus monkey
