Assay Details
Amplicon Length: | 92 |
Chromosome Location: | Chr. 1 - 122309801 - 122309801 [+] on 122309801 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-344a-5p | MIMAT0004654 |
| Rat | rno-miR-344a-3p | MIMAT0000592 |
Stem-loop Details
Rat

Genomic Map