Assay Details
Amplicon Length: | 146 |
Chromosome Location: | Chr. X - 118190532 - 118190532 [+] on 118190532 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-1298 | MIMAT0025060 |
Stem-loop Details
Rat
Please enter the information below and press OK to send your cart to Core Services for purchase.

Amplicon Length: | 146 |
Chromosome Location: | Chr. X - 118190532 - 118190532 [+] on 118190532 Build Rnor_6.0 |
Species: | Rat |
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-1298 | MIMAT0025060 |