Assay Details
Gene Family ID: | MIPF0000598, mir-1298 |
Mature miRNA Sequence: | UUCAUUCGGCUGUCCAGAUGUA |
Mature miRNA Details:
| Species | miRBase ID | miRBase Accession Number | |
|---|---|---|---|
| Human | hsa-miR-1298-5p | MIMAT0005800 | |
| Mouse | mmu-miR-1298-5p | MIMAT0014809 | |
| Rat | rno-miR-1298 | MIMAT0025060 | |
| Bovine | bta-miR-1298 | MIMAT0009970 | |
| Horse | eca-miR-1298 | MIMAT0013196 | |
| Pan troglodytes | ptr-miR-1298 | MIMAT0008017 | |
| View More Rows | |||
| Pongo pygmaeus | ppy-miR-1298 | MIMAT0016219 | |
| Rabbit | ocu-miR-1298-5p | MIMAT0048595 | |
| Rhesus monkey | mml-miR-1298-5p | MIMAT0024316 | |
Stem-loop Details
Human
Mouse
Rat
Bovine
Horse
Pan troglodytes
Pongo pygmaeus
Rabbit
Rhesus monkey
More Information
Related Products
miRBase Alias:
Human: hsa-miR-1298 (v19)
Mouse: mmu-miR-1298 (v17)
Macaca mulatta: mml-miR-1298 (v19)
Mouse: mmu-miR-1298 (v17)
Macaca mulatta: mml-miR-1298 (v19)

Genomic Map