Assay Details
Gene Family ID: | MIPF0000598, mir-1298 |
Mature miRNA Sequence: | UUCAUUCGGCUGUCCAGAUGUA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Human | hsa-miR-1298-5p | MIMAT0005800 |
| Mouse | mmu-miR-1298-5p | MIMAT0014809 |
| Rat | rno-miR-1298 | MIMAT0025060 |
| Bovine | bta-miR-1298 | MIMAT0009970 |
| Horse | eca-miR-1298 | MIMAT0013196 |
| Pan troglodytes | ptr-miR-1298 | MIMAT0008017 |
| View More Rows | ||
| Pongo pygmaeus | ppy-miR-1298 | MIMAT0016219 |
| Rabbit | ocu-miR-1298-5p | MIMAT0048595 |
| Rhesus monkey | mml-miR-1298-5p | MIMAT0024316 |
Stem-loop Details
Human
Mouse
Rat
Bovine
Horse
Pan troglodytes
Pongo pygmaeus
Rabbit
Rhesus monkey

Genomic Map