Assay Details
Gene Family ID: | MIPF0000390, mir-873 |
Mature miRNA Sequence: | GAGACUGACAAGUUCCCGGGA |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-873a-3p | MIMAT0017279 |
| Rat | rno-miR-873-3p | MIMAT0017318 |
Stem-loop Details
Mouse
Rat

Genomic Map