Assay Details
Amplicon Length: | 85 |
Chromosome Location: | Chr. 4 - 36668303 - 36668303 [-] on 36668303 Build GRCm38 |
Species: | Mouse |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Mouse | mmu-miR-873a-3p | MIMAT0017279 |
| Mouse | mmu-miR-873a-5p | MIMAT0004936 |
| Mouse | mmu-miR-873b | MIMAT0025177 |
Stem-loop Details
Mouse

Genomic Map