Assay Details
Amplicon Length: | 87 |
Chromosome Location: | Chr. 5 - 52481329 - 52481329 [+] on 52481329 Build Rnor_6.0 |
Species: | Rat |
miRBase Details:
| Species | miRBase ID | miRBase Accession Number |
|---|---|---|
| Rat | rno-miR-873-3p | MIMAT0017318 |
| Rat | rno-miR-873-5p | MIMAT0005339 |
Stem-loop Details
Rat

Genomic Map