Learn More
Copan Diagnostics ESwab™ Patented Sample Collection and Delivery System, Non Capture Cap
SDP
Catalog No. 23-600-905 Shop All Copan Diagnostics Inc. Products
Change view
Click to view available options
Sterility:
Sterile
Catalog No. Sterility
23-600-905 Sterile
1 options
This item is not returnable. View return policy
Catalog No. 23-600-905 Supplier Copan Diagnostics Supplier No. 480CFA
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy
Liquid based collection and transport device for aerobic, anaerobic, and fastidious bacteria for up to 48 hours. Polypropylene NON CAPTURE screw-cap tube filled with 1mL of Liquid Amies medium, plus one regular size Nylon flocked swab.
  • Maintains viability of aerobic, anaerobic, and fastidious bacteria for up to 48 hours
  • Using the patented FLOQSwab technology ESwab elutes the patient sample upon contact with the medium
  • Better growth and pathogen recovery - elutes entire sample upon contact with transport media
  • Equivalent sample distribution across all media plates
  • Better gram slides - homogenous preparation without specimen debris
  • Simplify and streamline - consolidate less optimal collection swabs into one superior system
  • One swab for up to 10 tests
  • Allows for easy automated specimen processing
  • Open Platform - Traditional bacteria, automation, gram stain, molecular assays, rapid antigen tests
  • Note: Product does not have capture cap
Specifications
Tip Material Flocked Nylon
Includes Single Regular Size Nylon Flocked Swab with 80 mm Breakpoint packaged with 1 mL Liquid Amies Medium in Self-Standing Tube with Plastic Purple Cap
Sterility Sterile
For Use With (Application) Collection and Transport System for Microbiology Swab Samples
Cap Color Purple
Media Type Liquid Amies Medium
Breakpoint 80 mm