Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ KingFisher Apex Purification Systems
GSA_VA

Catalog No. 54-009-20
Encompass
Change view
Click to view available options
Product Type:
24 Combi head
96 Combi head
96 DW head
96 PCR Head
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
54-009-20 96 Combi head
54-009-10 96 PCR Head
54-009-30 96 DW head
54-009-40 24 Combi head
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 54-009-20 Supplier Thermo Scientific™ Supplier No. 5400920

This product is not available for sale online. Please contact your Life Science Specialist for assistance with purchasing.

The KingFisher Apex Purification System is a versatile and user-friendly instrument that uses magnetic particle processing to automate the purification or isolation of DNA, RNA, proteins, and cells from virtually any source.

The KingFisher Apex Purification System is a versatile and user-friendly instrument that uses magnetic particle processing to automate the purification or isolation of DNA, RNA, proteins, and cells from virtually any source. It moves magnetic beads, not liquids, through the main steps of sample preparation: bind, wash, and elute. This process provides excellent quality, reproducible results, and reduces hands-on time for sample processing.

  • User-friendly high throughput sample purification system for automating DNA/RNA, protein and cell purification
  • Provides versatile and high-speed purification with reproducible results
  • Easy to use interface with touch screen empowering the user to control all details of the application and purification process
  • BindIx PC Software for added convenience
  • Extensive selection of validated protocols for different type of applications
  • Cloud library for quick protocol retrieval

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type 96 Combi head
Sample Type Plasma, Blood, Urine, Saliva, Cell Homogenate, Tissue Homogenate
Suitable Downstream Analysis PCR/qPCR, Sequencing, Mass Spec
Volume (Metric) Processing KF 96 plate 15 to 200 μL, 96 DW plate 15 to 1000 μL, 96 storage tube 30 to 200 μL
Plates per Deck 8
Format 96-well
Capacity 96 Samples/Run
Collection Efficiency of Particles >98% with 2.8 μm particle size
Cooling Temperature Down to +4°C
Display Touch screen (10 in.)
For Use With (Application) Genomics and proteomics, target identification, veterinary assays, biomarker discovery, quality control, forensics
Heating Temperature +4°C above ambient temperature up to +100°C
Magnetic Rods 96
Particle Size (Metric) ca. >1 μm
Samples per Run 96
Type of Magnet Head 96 in one frame, interchangeable
UV Exposure Time Max 23 hr. 59 min.
UV Lamp 2 pcs
Plate Type 96 KF, 96 DW, 96 storeage tube
Depth (English) 22 in.
Depth (Metric) 56 cm
Height (English) 18 in.
Height (Metric) 46 cm
Weight (English) 123 lb.
Weight (Metric) 56 kg
Width (English) 31 in.
Width (Metric) 79 cm
Show More Show Less
What should I do if the tip comb does not load correctly onto my Thermo Scientific KingFisher system?

If the tip does not load correctly onto a Thermo Scientific KingFisher magnetic head, we recommend that you try manually flexing the tips both lengthwise and widthwise to level the tip comb. A video of flexing the tip comb can be found here: https://www.thermofisher.com/us/en/home/life-science/dna-rna-purification-analysis/automated-purification-extraction/kingfisher-systems/resources.html. The video is under "KingFisher Flex" and titled "How to install the magnet head and check the tip comb." Flexing of the tip comb is demonstrated at timepoint 1:35. If that does not work, try a different box or lot of tips. This applies to all KingFisher instruments.

WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.