Learn More
CPC Scientific Ac-Nle-cyclo(Asp-His-DPhe-Arg-Trp-Lys-NH2)
(Peptide beta bond Asp3 and Lys8 bridge) 2MG
SDP
Catalog No. 502110424
Encompass
Cyclic MSH analog is a potent full agonist of the melanocortin-3 and melanocortin-4 receptors (MC3-R and MC4-R). MTII is a useful tool in the study of the agouti obesity syndrome and the role of melanocortinergic neurons in feeding.SEQUENCE: Ac-Nle-cyclo(Asp-His-DPhe-Arg-Trp-Lys-NH2)(Peptide beta bond Asp3 and Lys8 bridge)ONE-LETTER SEQUENCE: Ac-Nle-cyclo(DHfRWK-NH2)(Peptide beta bond D3 and K8 bridge)MOLECULAR FORMULA: C50H69N15O9MOLECULAR WEIGHT: 1024.19STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [121062-08-6]SYNONYMS: MTII, Bremelanotide amide, Melanotan IIRESEARCH AREA: GastrointestinalREFERENCES:V.J. Hruby et al., J. Med. Chem., 38, 3435 (1995)
SKU(s): SOMA-003A, SOMA-003B
Catalog No. 50-211-0424 Supplier CPC Scientific Supplier No. SOMA003B
Add to Cart
Add to Cart
Cyclic MSH analog is a potent full agonist of the melanocortin-3 and melanocortin-4 receptors (MC3-R and MC4-R). MTII is a useful tool in the study of the agouti obesity syndrome and the role of melanocortinergic neurons in feeding.SEQUENCE: Ac-Nle-cyclo(Asp-His-DPhe-Arg-Trp-Lys-NH2)(Peptide beta bond Asp3 and Lys8 bridge)ONE-LETTER SEQUENCE: Ac-Nle-cyclo(DHfRWK-NH2)(Peptide beta bond D3 and K8 bridge)MOLECULAR FORMULA: C50H69N15O9MOLECULAR WEIGHT: 1024.19STORAGE CONDITIONS: -20 5 CCAS REGISTRY NUMBER: [121062-08-6]SYNONYMS: MTII, Bremelanotide amide, Melanotan IIRESEARCH AREA: GastrointestinalREFERENCES:V.J. Hruby et al., J. Med. Chem., 38, 3435 (1995)
SKU(s): SOMA-003A, SOMA-003B