Learn More
ACROBiosystems Chemiluminescent Substrate Solution (HRP Marker)
Supplier:  ACROBiosystems ABK002200ML
Encompass_Preferred
The Chemiluminescent Substrate Solution (HRP Marker) is a specific triggering reagent for peroxidase labels such as horseradish peroxidase (HRP) and can be used in ELISA procedures. This substrate is an enhanced chemiluminescent substrate enables detection at femtogram range of target by oxidizing luminol in the presence of horseradish peroxidase (HRP) and peroxide.This product is provided in two liquid buffer of Solution A (Luminol-Based Solution) and Solution B (Peroxide Solution). When used, mix Solution A and Solution B in equal volume by 1:1, and it gives a rapid light emission.
Catalog No. 50-233-1104
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.