Learn More
Hach Company Conductivity Standard Solution, 146.9 µS/cm, KCl, 500 mL
Catalog No. 501996981 Shop All Hach Company Products
Encompass
Conductivity Standard Solution. 146.9 µS/cm ±10% at 25°C, 74.55 mg/L ±10% as Potassium Chloride (KCl) (0.001M). 500 mL bottle.
Catalog No. 50-199-6981 Supplier Hach Company Supplier No. 2974249
Conductivity Standard Solution. 146.9 µS/cm ±10% at 25°C, 74.55 mg/L ±10% as Potassium Chloride (KCl) (0.001M). 500 mL bottle.