Learn More
ONLINE METALS 0.003" ALUMINUM FOIL 1000 SERI
Catalog No. NC1924740
Encompass
Aluminum Foil 1145/1235/1100 O Temper is mill-finished and is an excellent conductor of heat and electricity. This aluminum foil meets ASTM B479 and QQ-A-1876.
Catalog No. NC1924740 Supplier ONLINE METALS Supplier No. 24342
Aluminum Foil 1145/1235/1100 O Temper is mill-finished and is an excellent conductor of heat and electricity. This aluminum foil meets ASTM B479 and QQ-A-1876.