Learn More
Analytical Sales & Services 0.005" PFA Fin Separator Channel Films
Catalog No. 5000805134
Encompass
PFA Fin Separator Channel Films\nfor use with Flow Electrolysis Cells\n\nThickness : .005”  (Ideal for users looking to further reduce the inter-electrode gap)\nColor: clear\n\nQty/Pkg: 25\n\n \n\n 
Catalog No. 50-008-05134 Supplier Analytical Sales & Services Supplier No. F1088710
PFA Fin Separator Channel Films\nfor use with Flow Electrolysis Cells\n\nThickness : .005”  (Ideal for users looking to further reduce the inter-electrode gap)\nColor: clear\n\nQty/Pkg: 25\n\n \n\n 