Learn More
Analytical Sales & Services 0.005" PFA Isolation Layer Films
Catalog No. 5000805111
Encompass
PFA Isolation Layer Films\nfor use with Flow Electrolysis Cells\n\nThickness : .005”  (Ideal for users looking to further reduce the inter-electrode gap)\nColor: clear\n\nQty/Pkg: 25
Catalog No. 50-008-05111 Supplier Analytical Sales & Services Supplier No. F1088705
PFA Isolation Layer Films\nfor use with Flow Electrolysis Cells\n\nThickness : .005”  (Ideal for users looking to further reduce the inter-electrode gap)\nColor: clear\n\nQty/Pkg: 25