Learn More
Analytical Sales & Services 0.005" PFA Tangential Mixer Channel Films
Catalog No. 5000805100
Encompass
PFA Tangential Mixer Channel Films\nfor use with Flow Electrolysis Cells\n\nThickness : .005”  (Ideal for users looking to further reduce the inter-electrode gap)\nColor: clear\n\nQty/Pkg: 25
Catalog No. 50-008-05100 Supplier Analytical Sales & Services Supplier No. F1088707
PFA Tangential Mixer Channel Films\nfor use with Flow Electrolysis Cells\n\nThickness : .005”  (Ideal for users looking to further reduce the inter-electrode gap)\nColor: clear\n\nQty/Pkg: 25