Learn More
Analytical Sales & Services 0.010" PFA Flooded Flow Channel Films
Catalog No. 5000805376
Encompass
PFA Flooded Flow Channel Films\nfor use with Flow Electrolysis Cells\n\n• Thickness: .010”\n• Color: clear\n• Use to achieve maximum active surface area\n• For reactions benefitting from an increased active surface area, achieves up to 2x yield vs our meandering channel\n\nQty/Pkg: 25
Catalog No. 50-008-05376 Supplier Analytical Sales & Services Supplier No. F1088412
Add to Cart
Add to Cart
PFA Flooded Flow Channel Films\nfor use with Flow Electrolysis Cells\n\n• Thickness: .010”\n• Color: clear\n• Use to achieve maximum active surface area\n• For reactions benefitting from an increased active surface area, achieves up to 2x yield vs our meandering channel\n\nQty/Pkg: 25