Learn More
Analytical Sales & Services 0.010" PFA Meandering Channel Films
Catalog No. 5000805096
Encompass
PFA Meandering Channel Films\nfor use with Flow Electrolysis Cells\n\nThickness : .010”\nColor: clear\n\nQty/Pkg: 25
Catalog No. 50-008-05096 Supplier Analytical Sales & Services Supplier No. F1088403
PFA Meandering Channel Films\nfor use with Flow Electrolysis Cells\n\nThickness : .010”\nColor: clear\n\nQty/Pkg: 25