Learn More
Hach Company Conductivity Standard Solution, 1412 µS/cm, KCl, 50 mL
Supplier:  Hach Company 2974326
Encompass
Conductivity Standard Solution. 1412 µS/cm ±10% at 25°C, 745.5 mg/L ±10% as Potassium Chloride (KCl) (0.01M). 50 mL pour-top bottle.
Catalog No. 50-199-6982