Learn More
Glen Research 0.02M IODINE IN TETRAHY2,000ML
Catalog No. NC2219397
Encompass_Preferred
0.02M Iodine in Tetrahydrofuran/Pyridine/Water (70:20:10) 2,000mL
This item is not returnable. View return policy
Catalog No. NC2219397 Supplier Glen Research Supplier No. 40433062
0.02M Iodine in Tetrahydrofuran/Pyridine/Water (70:20:10) 2,000mL