Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aqua Solutions Potassium Hydroxide 0.02 Normal In Methanol N.i.s.t. Traceable 4L
SDP

Catalog No. NC9682847
Encompass_Preferred

Potassium Hydroxide 0.02 Normal In Methanol N.i.s.t. Traceable 4L

Catalog No. NC9682847 Supplier Aqua Solutions Supplier No. 72394L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Potassium Hydroxide 0.02 Normal In Methanol N.i.s.t. Traceable 4L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.