Learn More
Charles River Laboratories 0.03 Lysate 50-test vial (5.2 mL), 12 Pack
Catalog No. NC9236518
Encompass
LYSATE Limulus Amebocyte Lysate, Gel Clot Reagent, This Lysate Forms a Firm Gel-Clot for More Accurate Results and is Buffered for Better Interference Resistance.Volume: 5.2mL
Catalog No. NC9236518 Supplier Charles River Laboratories Supplier No. R15003/DEL
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
LYSATE Limulus Amebocyte Lysate, Gel Clot Reagent, This Lysate Forms a Firm Gel-Clot for More Accurate Results and is Buffered for Better Interference Resistance.Volume: 5.2mL