Learn More
Waters Corp Conversion Kit, 0.045, Seal Wash Housing, Seals
Catalog No. 502391979 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This conversion kit is an ACQUITY I-Class BSM 0.045 kit that includes a Seal Wash Housing with Seals. It is designed as an upgrade kit for ACQUITY UPLC systems.
  • Contains seal wash housing with seals
  • Designed for UPLC systems
  • Compatible with UPLC H-Class system
  • Replacement is on an as-required basis
Catalog No. 50-239-1979 Supplier Waters Corp Supplier No. 205001097
This conversion kit is an ACQUITY I-Class BSM 0.045 kit that includes a Seal Wash Housing with Seals. It is designed as an upgrade kit for ACQUITY UPLC systems.
  • Contains seal wash housing with seals
  • Designed for UPLC systems
  • Compatible with UPLC H-Class system
  • Replacement is on an as-required basis