Learn More
Hach Company Certified Conductivity Standard Solution, 1015 µS/cm, 0.05% NaCl, 500 mL
Catalog No. 501998103 Shop All Hach Company Products
Encompass
Certified Conductivity Standard Solution with Certificate of Conformity and Traceability* reporting exact conductivity value with related expanded uncertainty (k=2). Guaranteed shelf-life thanks to premium aluminium packaging. 0.05% Sodium Chloride (NaCl) Conductivity: 1015 µS/cm ±0.5% (k=2) at 25°C, 500 mL bottle.
Catalog No. 50-199-8103 Supplier Hach Company Supplier No. S51M004
Certified Conductivity Standard Solution with Certificate of Conformity and Traceability* reporting exact conductivity value with related expanded uncertainty (k=2). Guaranteed shelf-life thanks to premium aluminium packaging. 0.05% Sodium Chloride (NaCl) Conductivity: 1015 µS/cm ±0.5% (k=2) at 25°C, 500 mL bottle.