Learn More
Chata Biosystems Sodium Phosphate, 0.05M, pH 6.8 - 100L CHEM+NECT
Catalog No. NC9434731
Encompass
Our CHEM+NECT™ system is designed to provide a closed system delivery option of introducting reagents into a clean and microbiological-controlled process. Designed to provide a closed system delivery option of introducing reagents into a clean and microbiological-controlled process. The CHEM+NECTTM bag is a sanitary and more environmentally sustainable alternative to glass and plastic bottles. The CHEM+NECTTM bag is ideal for processes in which pumping minor volumes of reagents and chemicals is used.
Catalog No. NC9434731 Supplier Chata Biosystems Supplier No. 56468
Our CHEM+NECT™ system is designed to provide a closed system delivery option of introducting reagents into a clean and microbiological-controlled process. Designed to provide a closed system delivery option of introducing reagents into a clean and microbiological-controlled process. The CHEM+NECTTM bag is a sanitary and more environmentally sustainable alternative to glass and plastic bottles. The CHEM+NECTTM bag is ideal for processes in which pumping minor volumes of reagents and chemicals is used.