Learn More
Charles River Laboratories 0.06 EU/ML LYSATE FOR GEL CLOT
Catalog No. NC2228986
Encompass
Lysate for Gel clot assay. ..
This item is not returnable. View return policy
Catalog No. NC2228986 Supplier Charles River Laboratories Supplier No. R15006
Lysate for Gel clot assay. ..