Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

0.07% n-Hexylamine in HPLC Grade Ethyl Acetate, Reagents

Catalog No. 01334695
Change view
Click to view available options
:
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-334-695 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-695 Supplier Reagents Holdings Llc Supplier No. CN0006004C
Only null left
Add to Cart
Add to Cart

0.07% n-Hexylamine in HPLC Grade Ethyl Acetate, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 141-78-6 , 111-26-2
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
InChI Key XEKOWRVHYACXOJ-UHFFFAOYSA-N
Synonym n-Hexylamine, 1-Aminohexane
IUPAC Name ethyl acetate
SMILES CCOC(C)=O

Specifications

Color Colorless
Boiling Point 77°C
Packaging Glass Bottle
Quantity 4 L
Specific Gravity 0.9009
Grade Reagent
Physical Form Liquid
Chemical Name or Material 0.07% n-Hexylamine in HPLC Grade Ethyl Acetate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.