Learn More
USA Scientific Inc 10 ul XL TipOne natural tip starter system
Supplier:  USA Scientific Inc 11103500
Encompass
TipOne 10 ul XL natural ultra micro pipet tip starter system, 10 racks and 2 packs of refill wafers (2880 tips)
Catalog No. NC1393058