Learn More
Metrohm USA 0.1 M Silver Nitrate Solution, 6 x 1 L bottles
SDP
Catalog No. NC0451835 Shop All Metrohm USA Products
Encompass
The Metrohm USA 0.1 M Silver Nitrate solution is a carefully formulated titrant designed for analytical applications. It is provided in a case containing six 1-liter bottles, making it convenient for laboratory use. The solution offers high purity and accuracy for precise measurement in volumetric titrations, allowing for the reliable determination of halides, sulfides, and other analytes. Its ready-to-use formulation saves time and enhances workflow efficiency in various scientific settings.
  • High purity and accuracy for critical analytical applications.
  • Versatile titrant for precipitation titrations across multiple analytes.
  • Convenient packaging of six 1-liter bottles for reduced waste.
  • Ready-to-use solution that eliminates preparation needs.
Catalog No. NC0451835 Supplier Metrohm USA Supplier No. LABTI92101
The Metrohm USA 0.1 M Silver Nitrate solution is a carefully formulated titrant designed for analytical applications. It is provided in a case containing six 1-liter bottles, making it convenient for laboratory use. The solution offers high purity and accuracy for precise measurement in volumetric titrations, allowing for the reliable determination of halides, sulfides, and other analytes. Its ready-to-use formulation saves time and enhances workflow efficiency in various scientific settings.
  • High purity and accuracy for critical analytical applications.
  • Versatile titrant for precipitation titrations across multiple analytes.
  • Convenient packaging of six 1-liter bottles for reduced waste.
  • Ready-to-use solution that eliminates preparation needs.