Learn More
Sigma Aldrich Fine Chemicals Biosciences 0.1 % FA IN WATER
Supplier:  Sigma Aldrich Fine Chemicals Biosciences 9006874X4L
Encompass
Water solutioncontains 0.1 % (v/v) formic acid, for UHPLC, for mass spectrometry
Catalog No. NC1647494