Learn More
Cell Biologics 0.1 % Gelatin-Based Coating Solution- Ready to use, 100ml
SDP
Catalog No. NC2754481
Encompass_Preferred
0.1 % Gelatin-Based Coating Solution is designed to coat cell culture flasks or dishes. Coat sterile culture dishes or flasks with Gelatin-Based Coating Solution for 2 min and then aspirate the excess solution before seeding cells. The use of coated-flasks improves cell attachment for certain types of primary cells.Store the solution at 2-8°C.
Catalog No. NC2754481 Supplier Cell Biologics Supplier No. 6950
0.1 % Gelatin-Based Coating Solution is designed to coat cell culture flasks or dishes. Coat sterile culture dishes or flasks with Gelatin-Based Coating Solution for 2 min and then aspirate the excess solution before seeding cells. The use of coated-flasks improves cell attachment for certain types of primary cells.Store the solution at 2-8°C.