Learn More
STEMCELL Technologies 0.1% Gelatin in Water, Size: 500 mL
SDP
Catalog No. NC1620050
Encompass
This product is recommended for the coating of culture dishes or flasks to grow mouse embryonic stem cells (mESCs), mouse induced pluripotent stem cells (miPSCs) or primary embryonic fibroblasts (PEFs).
Catalog No. NC1620050 Supplier STEMCELL Technologies Supplier No. 07903
This product is recommended for the coating of culture dishes or flasks to grow mouse embryonic stem cells (mESCs), mouse induced pluripotent stem cells (miPSCs) or primary embryonic fibroblasts (PEFs).