Learn More
TRIBIOSCIENCE INC 0.1% Gelatin Solution, 500 ML
SDP
Catalog No. NC3919359
Encompass
0.1% Gelatin Solution can be used to coat cell culture flasks and plates. The use of coated flasks and plates improves cell attachment for certain types of primary cells as well as certain types of continuous cell lines. Applications include coating cell culture plates or flasks to improve cell attachment for a variety of cell types.
Catalog No. NC3919359 Supplier TRIBIOSCIENCE INC Supplier No. TBS8004500
0.1% Gelatin Solution can be used to coat cell culture flasks and plates. The use of coated flasks and plates improves cell attachment for certain types of primary cells as well as certain types of continuous cell lines. Applications include coating cell culture plates or flasks to improve cell attachment for a variety of cell types.