Learn More
ATCC 0.1% Gelatin Solution, 100ml, Sterile
Catalog No. 50189667FP
Encompass
0.1% Gelatin Solution is used to coat cell culture flasks. The use of coated flasks imporves cell attachment for certain tupes of primary cells as well as certain continuous cell lines. New ATCC customers are required to register with ATCC prior to purchase. The application can be found at https://www.atcc.org/en/My_Profile/Register.aspx
Catalog No. 50-189-667FP Supplier ATCC Supplier No. PCS999027
0.1% Gelatin Solution is used to coat cell culture flasks. The use of coated flasks imporves cell attachment for certain tupes of primary cells as well as certain continuous cell lines. New ATCC customers are required to register with ATCC prior to purchase. The application can be found at https://www.atcc.org/en/My_Profile/Register.aspx