Learn More
Complement Technology Inc MgEGTA, 0.1 M MgCl2, 0.1 M EGTA, 10 ml, pH 7.3, Frozen liquid, 0.22 μm filtered, Research Use Only
Catalog No. NC1038078
Encompass
MgEGTA (Magnesium EGTA) is a precisely formulated buffer solution designed for critical applications in alternative complement pathway assays. This product is a frozen liquid reagent composed of 0.1 M MgCl2 and 0.1 M EGTA, adjusted to a pH of 7.3. It is 0.22 μm filtered without preservatives, ensuring high purity and stability for sensitive research. Functions effectively to prevent complement activation via the classical and lectin pathways, making it an essential tool for studies requiring specific control over complement cascade activation.
  • Composed of 0.1 M MgCl2 and 0.1 M EGTA
  • Maintains a pH of 7.3
  • 0.22 μm filtered for high purity
  • Frozen liquid form enhances stability
  • No preservatives used
  • Storage temperature options: 4 °C or -20 °C or below
  • Manufactured in the USA
  • Intended for research use only
  • Not classified as a hazardous substance
Catalog No. NC1038078 Supplier Complement Technology Inc Supplier No. B106
MgEGTA (Magnesium EGTA) is a precisely formulated buffer solution designed for critical applications in alternative complement pathway assays. This product is a frozen liquid reagent composed of 0.1 M MgCl2 and 0.1 M EGTA, adjusted to a pH of 7.3. It is 0.22 μm filtered without preservatives, ensuring high purity and stability for sensitive research. Functions effectively to prevent complement activation via the classical and lectin pathways, making it an essential tool for studies requiring specific control over complement cascade activation.
  • Composed of 0.1 M MgCl2 and 0.1 M EGTA
  • Maintains a pH of 7.3
  • 0.22 μm filtered for high purity
  • Frozen liquid form enhances stability
  • No preservatives used
  • Storage temperature options: 4 °C or -20 °C or below
  • Manufactured in the USA
  • Intended for research use only
  • Not classified as a hazardous substance