Learn More
Thomas Scientific Glass Beads, 0.1-.15 mm Dia., 1 Lb.
SDP
Catalog No. NC0068057
Encompass
Glass has a density of 2.5 g/cc and is the most commonly used bead media for "beadbeating." Beads are sold in one pound bottles with easy pour spouts. Zirconia beads have a density of 5.5g/cc (Twice as dense as glass beads - good for tough tissue). Sold in one pound bottles. Listed bead sizes are median (average) values. Because beads are sorted by sieves, bead size varies between +/-10% of a listed value.
Catalog No. NC0068057 Supplier Thomas Scientific Supplier No. 3411F08
Glass has a density of 2.5 g/cc and is the most commonly used bead media for "beadbeating." Beads are sold in one pound bottles with easy pour spouts. Zirconia beads have a density of 5.5g/cc (Twice as dense as glass beads - good for tough tissue). Sold in one pound bottles. Listed bead sizes are median (average) values. Because beads are sorted by sieves, bead size varies between +/-10% of a listed value.