Learn More
BioSpec Products 0.1 MM Zirconia/Silica Beads
SDP
Catalog No. NC0362415
Encompass_Preferred
Zirconia/Silica has a density of 3.7g/cc (50% more dense than glass - good for spores and most tissues) Beads are sold in one pound bottles. 0.1mm diameter.
Catalog No. NC0362415 Supplier BioSpec Products Supplier No. 11079101Z
Zirconia/Silica has a density of 3.7g/cc (50% more dense than glass - good for spores and most tissues) Beads are sold in one pound bottles. 0.1mm diameter.