Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

0.1 N Sodium Acetate in Ethanol, Unstandardized, Reagents

Catalog No. 01333957
Change view
Click to view available options
Quantity: 20 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-333-957 Quantity: 20 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-333-957 Supplier Reagents Holdings Llc Supplier No. CS01230020E7
 Restricted Item
Add to Cart
Add to Cart

0.1 N Sodium Acetate in Ethanol, Unstandardized, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Boiling Point 78°C
Chemical Name or Material 0.1 N Sodium Acetate in Ethanol, Unstandardized
Color Colorless
CAS 64-17-5, 7732-18-5, 67-56-1
Physical Form Liquid
Quantity 20 L
Synonym Sodium acetate/Ethyl Alcohol
Grade Reagent
Packaging HDPE Winpak
Specific Gravity 0.8236
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.