Learn More
MOLTOX Molecular Toxicology Inc Peptone Broth 0.1%, 9mL/tube, 10 tubes/box, Store at Room Temperature
SDP
Catalog No. 50948704
Encompass_Preferred
Peptone Broth is used as an organic nitrogen source for the cultivation of a variety of bacteria and fungi. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.
Catalog No. 50-948-704 Supplier MOLTOX Molecular Toxicology Inc Supplier No. 22101
Peptone Broth is used as an organic nitrogen source for the cultivation of a variety of bacteria and fungi. Each lot of media from MOLTOX is tested for sterility, where applicable, and its ability to support the growth of the appropriate microorganism. All media are accompanied by GLP-compliant formulation and Quality Control certificate.