Learn More
MSC Mitutoyo 293-831-30 0mm to 1" Carbide Electronic Outside Micrometer 0.0010mm Resolution, 0.000100" Accuracy, Ratchet Stop Thimble
Catalog No. 501113419
Encompass
Mitutoyo 293-831-30 0mm to 1" Carbide Electronic Outside Micrometer 0.0010mm Resolution, 0.000100" Accuracy, Ratchet Stop Thimble
Catalog No. 50-111-3419 Supplier MSC Supplier No. 04081394
Mitutoyo 293-831-30 0mm to 1" Carbide Electronic Outside Micrometer 0.0010mm Resolution, 0.000100" Accuracy, Ratchet Stop Thimble