Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

0.1% Trifluoroacetic Acid in Water, Solstice

Catalog No. 60090015 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
19 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-090-015 19 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-015 Supplier Solstice Supplier No. WP48519P
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

LabReady™ Solvent Blend, Custom Reagent

TRUSTED_SUSTAINABILITY

Specifications

CAS 7732-18-5, 76-05-1, 7732-18-5
CAS Max % 99.90%, 0.10%, ≤100.00%
Quantity 19 L
CAS Min % ≥60.00%
Packaging Winpak
Physical Form Liquid
Purity Grade Notes HPLC grade blends
Boiling Point 100°C
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.