Learn More
Cytiva Acrodisc Syringe Filters with Supor Membrane, Sterile - 0.1 µm, 32 mm (50/pkg)
Supplier:  Cytiva 4651
Encompass
Acrodisc syringe filters with Supor membrane have superior performance in a variety of sterile laboratory filtration applications. The inherently hydrophilic Supor (polyethersulfone) membrane is low protein binding and provides fast filtration and high throughput.The sterile Acrodisc syringe filters are individually packaged in blister packs and sterilized using gamma irradiation to eliminate potential or harmful extractables common with other sterilization methods such as EtO.The 0.1 μm pore size allows for the sterilization of small volumes of buffers, culture media, and additives. The use of 0.1 μm pore size can reduce mycoplasma contamination. Mycoplasma is a common contaminant of cell cultures, the presence of such a contaminant can have drastic effects on the structure and function of the infected host cells.For research use only. This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. NC1796378