Learn More
Pinnacle Technology Inc 0.10” EEG Mouse Screws
SDP
Catalog No. NC1682898
Encompass
0.10"""" stainless steel screws are offered for use with the 8201 headmount. Two 0.10"""" screws are implanted for anterior placement per headmount. For posterior electrode placement, 0.12” screws (8212) are available for facilitating the curvature of the skull. The screws come in a package of 8.
Catalog No. NC1682898 Supplier Pinnacle Technology Inc Supplier No. 8209
0.10"""" stainless steel screws are offered for use with the 8201 headmount. Two 0.10"""" screws are implanted for anterior placement per headmount. For posterior electrode placement, 0.12” screws (8212) are available for facilitating the curvature of the skull. The screws come in a package of 8.