Learn More
USA Scientific Inc 0.1-10 uL flat gel tip .37 mm
Catalog No. 502745276
Encompass
10 uL flat sequencing gel tip, 0.37 mm OD orifice, sterilized, 96 tips/rack, 2 racks/pack (192 tips)
Catalog No. 50-274-5276 Supplier USA Scientific Inc Supplier No. 10124610
10 uL flat sequencing gel tip, 0.37 mm OD orifice, sterilized, 96 tips/rack, 2 racks/pack (192 tips)