Learn More
Chemglass Life Sciences 12mm PTFE Stem Only, Perfluoro O-Rings
SDP
Catalog No. CG51145
Encompass_Preferred
12mm PTFE Stem Only, Perfluoro O-Rings. Replacement 12mm Chem-Cap stem and control knob for Chemglass cylindrical and spherical vessels. PTFE stem is sold complete with Perfluoro or Viton o-rings installed under each Chem-Cap and one installed as the back up o-ring.
Catalog No. CG51145 Supplier Chemglass Life Sciences Supplier No. CG51145
12mm PTFE Stem Only, Perfluoro O-Rings. Replacement 12mm Chem-Cap stem and control knob for Chemglass cylindrical and spherical vessels. PTFE stem is sold complete with Perfluoro or Viton o-rings installed under each Chem-Cap and one installed as the back up o-ring.