Learn More
Accuflow Systems Saint-Gobain BarbLock Retainer, PVDF, 1/16 in ID, AL Series
Catalog No. NC3592552
Encompass_Preferred
The AL-series BarbLock PVDF one-piece retainer secures flexible tubing to barbed fittings for leak-resistant connections in bioprocessing and other critical fluid-transfer applications. The design provides a full 360° radial crimp behind the barb and 360° compression over the barb ridge to address leak-path issues, and PVDF construction offers chemical resistance and gamma stability.
Catalog No. NC3592552 Supplier Accuflow Systems Supplier No. BL135110ALPVDF
The AL-series BarbLock PVDF one-piece retainer secures flexible tubing to barbed fittings for leak-resistant connections in bioprocessing and other critical fluid-transfer applications. The design provides a full 360° radial crimp behind the barb and 360° compression over the barb ridge to address leak-path issues, and PVDF construction offers chemical resistance and gamma stability.