Learn More
Chemglass Life Sciences Valve, 0-12mm, 1-Arm, Glass Only
SDP
Catalog No. 501532011
Encompass_Preferred
Valve, 0-12mm, 1-Arm, Glass Only. This style hi-vac valve has a single sidearm seal at a 90 angle to the barrel. This inexpensive configuration allows for the lower portion of the barrel to be sealed directly to the apparatus being constructed, making for more compact, stronger assemblies. Designed with a precision external thread for fine control.
Catalog No. 50-153-2011 Supplier Chemglass Life Sciences Supplier No. CGK50203
Valve, 0-12mm, 1-Arm, Glass Only. This style hi-vac valve has a single sidearm seal at a 90 angle to the barrel. This inexpensive configuration allows for the lower portion of the barrel to be sealed directly to the apparatus being constructed, making for more compact, stronger assemblies. Designed with a precision external thread for fine control.